Learn More
Biolegend Alexa Fluor 488 anti-mouse CD38
Supplier:  Biolegend 102714
Encompass_Preferred
Alexa Fluor 488 anti-mouse CD38 [90]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-162-727