Learn More
Biolegend Alexa Fluor 488 anti-mouse CD4
Supplier:  Biolegend 100529
Encompass_Preferred
Alexa Fluor 488 anti-mouse CD4 [RM4-5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 100 ug
Catalog No. NC9066195