Learn More
Biolegend Alexa Fluor 488 anti-mouse CD4
Supplier:  Biolegend 100423
Encompass_Preferred
Alexa Fluor 488 anti-mouse CD4 [GK1.5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IHC-F, ICC; Size: 100 ug
Catalog No. NC9038069