Learn More
Biolegend Alexa Fluor 488 anti-mouse CD45
Supplier:  Biolegend 103122
Encompass_Preferred
Alexa Fluor 488 anti-mouse CD45 [30-F11]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC9900679