Learn More
Biolegend Alexa Fluor 488 anti-mouse CD90.2
Supplier:  Biolegend 105316
Encompass_Preferred
Alexa Fluor 488 anti-mouse CD90.2 [30-H12]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 100 ug
Catalog No. 50-604-852