Learn More
Biolegend Alexa Fluor 488 anti-MU/HU GL7 Antigen (T/B Cell Act. Marker)
Supplier:  Biolegend 144611
Encompass_Preferred
Alexa Fluor 488 anti-MU/HU GL7 Antigen (T/B Cell Act. Marker) [GL7]; Isotype: Rat IgM, κ; Reactivity: Mouse, Human; Apps: FC, IHC-F; Size: 25 ug
Catalog No. 50-711-896