Learn More
Biolegend Alexa Fluor 488 anti-mouse/human CD15 (SSEA-1)
Supplier:  Biolegend 125609
Encompass_Preferred
Alexa Fluor 488 anti-mouse/human CD15 (SSEA-1) [MC-480]; Isotype: Mouse IgM, κ; Reactivity: Mouse, Cross-Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-164-266