Learn More
Biolegend Alexa Fluor 488 anti-mouse IL-17A
Supplier:  Biolegend 506909
Encompass_Preferred
Alexa Fluor 488 anti-mouse IL-17A [TC11-18H10.1]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 25 ug
Catalog No. 50-170-000