Learn More
Biolegend Alexa Fluor 488 anti-mouse IL-4
Supplier:  Biolegend 504111
Encompass_Preferred
Alexa Fluor 488 anti-mouse IL-4 [11B11]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 25 ug
Catalog No. 50-169-833