Learn More
Biolegend Alexa Fluor 488 anti-mouse/rat CD29
Supplier:  Biolegend 102212
Encompass_Preferred
Alexa Fluor 488 anti-mouse/rat CD29 [HMβ1-1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Rat; Apps: FC, IHC-F; Size: 100 ug
Catalog No. 50-162-660