Learn More
Biolegend Alexa Fluor 488 anti-rat CD3
Supplier:  Biolegend 201406
Encompass_Preferred
Alexa Fluor 488 anti-rat CD3 [1F4]; Isotype: Mouse IgM, κ; Reactivity: Rat; Apps: FC, IHC-F; Size: 100 ug
Catalog No. 50-207-2476