Learn More
Biolegend Alexa Fluor 488 anti-rat IgG2b
Supplier:  Biolegend 408211
Encompass_Preferred
Alexa Fluor 488 anti-rat IgG2b [MRG2b-85]; Isotype: Mouse IgG1, κ; Reactivity: Rat; Apps: FC, IHC-F; Size: 100 ug
Catalog No. 50-207-9831