Learn More
Biolegend Alexa Fluor 488 anti-rat IgM
Supplier:  Biolegend 408908
Encompass_Preferred
Alexa Fluor 488 anti-rat IgM [MRM-47]; Isotype: Mouse IgG; Reactivity: Rat; Apps: FC, IHC-F; Size: 100 ug
Catalog No. 50-207-9850