Learn More
Biolegend Alexa Fluor 488 anti-RNA Polymerase II RPB1
Supplier:  Biolegend 664916
Encompass_Preferred
Alexa Fluor 488 anti-RNA Polymerase II RPB1 [8WG16]; Isotype: Mouse IgG2a; Reactivity: Human, Yeast, Wheat, Mouse, C. elegans, X. laevis; Apps: ICC; Size: 100 ug
Catalog No. 50-208-7226