Learn More
Biolegend Alexa Fluor 488 anti-STAT6 Phospho (Tyr641)
Supplier:  Biolegend 686007
Encompass_Preferred
Alexa Fluor 488 anti-STAT6 Phospho (Tyr641) [A15137E]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests
Catalog No. 50-208-7512