Learn More
Biolegend Alexa Fluor 488 anti-Tubulin Beta 3 (TUBB3)
Supplier:  Biolegend 657404
Encompass_Preferred
Alexa Fluor 488 anti-Tubulin Beta 3 (TUBB3) [AA10]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse, Rat; Apps: ICFC, IHC-F, 3-D IHC; Size: 100 ug
Catalog No. 50-605-930