Learn More
Biolegend Alexa Fluor 488 Donkey anti-rabbit IgG (min. x-reactivity)
Catalog No. 50712245
Encompass_Preferred
Alexa Fluor 488 Donkey anti-rabbit IgG (min. x-reactivity) [Poly4064]; Isotype: Donkey Polyclonal Ig; Reactivity: Rabbit; Apps: FC; Size: 100 ug
Catalog No. 50-712-245 Supplier Biolegend Supplier No. 406416
Alexa Fluor 488 Donkey anti-rabbit IgG (min. x-reactivity) [Poly4064]; Isotype: Donkey Polyclonal Ig; Reactivity: Rabbit; Apps: FC; Size: 100 ug