Learn More
Aviva Systems Biology Corp A4GALT Antibody - middle region (ARP47303_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47303_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against A4GALT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human A4GALT. Alternate names: A14GALT, A4GALT1, P1, PK, Gb3S, P(k)ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59514