Learn More
MSC MAXICUT ULTRA NTRL CR GLV
Supplier: MSC 08298317
Cut & Puncture Resistant Gloves; Coating Material: Micro-Foam Nitrile ; Coating Coverage: Palm & Fingers ; Glove Type: Cut-Resistant ; ANSI/ISEA Cut Resistance Level: A5 ; Primary Material: Engineered Yarn ; Lining Material: Engineered Yarn