Learn More
Invivogen A549-Dual Cells, 3-7 x 10e6 cells
SDP
Catalog No. NC1023701
Encompass
A549-Dual cells are adherent epithelial cells that have been derived from the human A549 lung carcinoma cell line by stable integration of two inducible reporter constructs. The A549 cell line is a well-characterized cellular model for asthma, allergies and respiratory infections.
Catalog No. NC1023701 Supplier Invivogen Supplier No. A549DNFIS
A549-Dual cells are adherent epithelial cells that have been derived from the human A549 lung carcinoma cell line by stable integration of two inducible reporter constructs. The A549 cell line is a well-characterized cellular model for asthma, allergies and respiratory infections.