Learn More
Eurofins Discoverx InCELL Hunter A549 G9a Methyltransferase Cell Line, 2 vials. (96-0003C15)
Supplier:  Eurofins Discoverx 960003C15
Encompass
The InCELL Hunter A549 G9a Methyltransferase Cell Line is a stable, engineered cell line for use in studying small molecule drug candidates that engage with regulators of epigenetics pathways. This cell line enables assessment of compound-engagement-based stabilization of G9a via detection of G9a levels. This product uses EFC technology. The cell line included overexpresses G9a target protein fused to ProLabel. In the absence of compound that binds and stabilizes the target, this protein is degraded and no EFC signal is produced. When compound engages and stabilizes the target, EA contained in the detection reagent produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/epigenetics.
Catalog No. 50-200-4127
Only null left
Add to Cart
Request a Quote