Learn More
Eurofins Discoverx InCELL Hunter A549 PRMT3 Methyltransferase Cell Line, 2 vials. (96-0072C15)
Catalog No. 502004509
Encompass
The InCELL Hunter A549 PRMT3 Methyltransferase Cell Line is a stable, engineered cell line for use in studying small molecule drug candidates that engage with regulators of epigenetics pathways. This cell line enables assessment of compound-engagement-based stabilization of PRMT3 via detection of PRMT3 levels. This product uses EFC technology. The cell line included overexpresses PRMT3 target protein fused to ProLabel. In the absence of compound that binds and stabilizes the target, this protein is degraded and no EFC signal is produced. When compound engages and stabilizes the target, EA contained in the detection reagent produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/epigenetics.
Catalog No. 50-200-4509 Supplier Eurofins Discoverx Supplier No. 960072C15
Add to Cart
Add to Cart
Request a Quote
The InCELL Hunter A549 PRMT3 Methyltransferase Cell Line is a stable, engineered cell line for use in studying small molecule drug candidates that engage with regulators of epigenetics pathways. This cell line enables assessment of compound-engagement-based stabilization of PRMT3 via detection of PRMT3 levels. This product uses EFC technology. The cell line included overexpresses PRMT3 target protein fused to ProLabel. In the absence of compound that binds and stabilizes the target, this protein is degraded and no EFC signal is produced. When compound engages and stabilizes the target, EA contained in the detection reagent produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/epigenetics.