Learn More
Biolegend Alexa Fluor 555 Donkey anti-rabbit IgG (minimal x-reactivity)
Catalog No. 501030875
Encompass_Preferred
Alexa Fluor 555 Donkey anti-rabbit IgG (minimal x-reactivity) [Poly4064]; Isotype: Donkey Polyclonal Ig; Reactivity: Rabbit; Apps: ICC; Size: 100 ug
Catalog No. 50-103-0875 Supplier Biolegend Supplier No. 406412
Alexa Fluor 555 Donkey anti-rabbit IgG (minimal x-reactivity) [Poly4064]; Isotype: Donkey Polyclonal Ig; Reactivity: Rabbit; Apps: ICC; Size: 100 ug