Learn More
Biolegend Alexa Fluor 594 anti-ATG5
Supplier:  Biolegend 847405
Encompass_Preferred
Alexa Fluor 594 anti-ATG5 [177.19]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: ICC; Size: 25 ug
Catalog No. 50-207-4519