Learn More
Biolegend Alexa Fluor 594 anti-Cytokeratin (pan reactive)
Supplier:  Biolegend 628606
Encompass_Preferred
Alexa Fluor 594 anti-Cytokeratin (pan reactive) [C-11]; Isotype: Mouse IgG1, κ; Reactivity: Reacts with conserved epitope in all species tested. Recognizes cytokeratin 4, 5, 6, 8, 10, 13, and 18; Apps: ICC; Size: 100 ug
Catalog No. 50-208-6855