Learn More
Biolegend Alexa Fluor 594 anti-Cytokeratin 18
Supplier:  Biolegend 628406
Encompass_Preferred
Alexa Fluor 594 anti-Cytokeratin 18 [DA-7]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICC, IHC-P; Size: 100 ug
Catalog No. 50-208-6848