Learn More
Biolegend Alexa Fluor 594 anti-H2A.X
Supplier:  Biolegend 600204
Encompass_Preferred
Alexa Fluor 594 anti-H2A.X [W16171A]; Isotype: Rat IgG2a, κ; Reactivity: Human, Mouse; Apps: ICC; Size: 100 ug
Catalog No. 50-208-6613