Learn More
Biolegend Alexa Fluor 594 anti-Histone H3 Acetylated Lysine 9 (K9ac)
Supplier:  Biolegend 698403
Encompass_Preferred
Alexa Fluor 594 anti-Histone H3 Acetylated Lysine 9 (K9ac) [2G1F9]; Isotype: Rat IgG2a, κ; Reactivity: Human, Mouse; Apps: ICC; Size: 25 ug
Catalog No. 50-208-7715