Learn More
Biolegend Alexa Fluor 594 anti-MAP2
Supplier:  Biolegend 801803
Encompass_Preferred
Alexa Fluor 594 anti-MAP2 [SMI 52]; Isotype: Mouse IgG1, κ; Reactivity: Mouse, Rat; Apps: IHC-P; Size: 100 ug
Catalog No. 50-208-9868