Learn More
Biolegend Alexa Fluor 594 anti-mouse CD326 (Ep-CAM)
Supplier:  Biolegend 118222
Encompass_Preferred
Alexa Fluor 594 anti-mouse CD326 (Ep-CAM) [G8.8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICC, IHC-F, 3-D IHC; Size: 100 ug
Catalog No. 50-711-853