Learn More
Biolegend Alexa Fluor 594 anti-mouse CD47
Supplier:  Biolegend 127531
Encompass_Preferred
Alexa Fluor 594 anti-mouse CD47 [miap301]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: IHC-F; Size: 25 ug
Catalog No. 50-207-0814