Learn More
Biolegend Alexa Fluor 594 anti-mouse CD68
Supplier:  Biolegend 137020
Encompass_Preferred
Alexa Fluor 594 anti-mouse CD68 [FA-11]; Isotype: Rat IgG2a; Reactivity: Mouse; Apps: IHC-F; Size: 100 ug
Catalog No. 50-711-877