Learn More
Biolegend Alexa Fluor 594 anti-mouse CD8a
Supplier:  Biolegend 100758
Encompass_Preferred
Alexa Fluor 594 anti-mouse CD8a [53-6.7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: IHC-F, FC; Size: 100 ug
Catalog No. 50-711-807