Learn More
Biolegend Alexa Fluor 594 anti-mouse CD8b (Ly-3)
Supplier:  Biolegend 126636
Encompass_Preferred
Alexa Fluor 594 anti-mouse CD8b (Ly-3) [YTS156.7.7]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: IHC-F; Size: 100 ug
Catalog No. 50-207-0748