Learn More
Biolegend Alexa Fluor 594 anti-mouse F4/80
Supplier:  Biolegend 123140
Encompass_Preferred
Alexa Fluor 594 anti-mouse F4/80 [BM8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: IHC-F; Size: 100 ug
Catalog No. 50-711-858