Learn More
Biolegend Alexa Fluor 594 anti-mouse Galectin-9
Supplier:  Biolegend 137906
Encompass_Preferred
Alexa Fluor 594 anti-mouse Galectin-9 [108A2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: IHC-F; Size: 100 ug
Catalog No. 50-207-1228