Learn More
Biolegend Alexa Fluor 594 anti NF-H/NF-M, Hypophosphorylated
Supplier:  Biolegend 835609
Encompass_Preferred
Alexa Fluor 594 anti NF-H/NF-M, Hypophosphorylated [SMI 35]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P, ICC; Size: 25 ug
Catalog No. 50-207-4337