Learn More
Biolegend Alexa Fluor 594 anti-Pax-5
Supplier:  Biolegend 649712
Encompass_Preferred
Alexa Fluor 594 anti-Pax-5 [1H9]; Isotype: Rat IgG2a, κ; Reactivity: Human, Mouse; Apps: ICC; Size: 100 ug
Catalog No. 50-208-6985