Learn More
Biolegend Alexa Fluor 594 anti-STAT3
Supplier:  Biolegend 678003
Encompass_Preferred
Alexa Fluor 594 anti-STAT3 [4G4B45]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: ICC; Size: 100 ug
Catalog No. 50-208-7386
Edge