Learn More
Biolegend Alexa Fluor 594 anti-α-Synuclein Phospho (Ser129)
Supplier:  Biolegend 825708
Encompass_Preferred
Alexa Fluor 594 anti-α-Synuclein Phospho (Ser129) [P-syn/81A]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: IHC-P; Size: 100 ug
Catalog No. 50-207-4226