Learn More
Biolegend Alexa Fluor 594 anti-TAL1
Supplier:  Biolegend 681004
Encompass_Preferred
Alexa Fluor 594 anti-TAL1 [4G8C07]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICC; Size: 100 ug
Catalog No. 50-208-7422