Learn More
MSC EDGE POLY/HPPE C/R GLV
Catalog No. 502737313
Encompass
Cut & Puncture Resistant Gloves; Style: Knit ; Coating Material: Polyurethane ; Coating Coverage: Palm ; Glove Type: Cut, Puncture & Abrasion-Resistant ; ANSI/ISEA Cut Resistance Level: A6 ; Primary Material: Polyester; Fiberglass; Spandex; Nylon; HPPE
Catalog No. 50-273-7313 Supplier MSC Supplier No. 74403155
Add to Cart
Add to Cart
Cut & Puncture Resistant Gloves; Style: Knit ; Coating Material: Polyurethane ; Coating Coverage: Palm ; Glove Type: Cut, Puncture & Abrasion-Resistant ; ANSI/ISEA Cut Resistance Level: A6 ; Primary Material: Polyester; Fiberglass; Spandex; Nylon; HPPE