Learn More
MSC Honeywell Gloves, Cut A6, Puncture 4, Abrasion 4, Nitrile, 13 Gauge
Catalog No. 5000508497
Encompass
These CoreShield gloves are designed for cut, puncture, and abrasion resistance, offering a lightweight and flexible solution for hand protection. They feature an engineered yarn and high-performance nitrile coating, providing comfort and dexterity for all-day wear in various industrial applications.
  • Engineered yarn for lightweight cut protection and dexterity.
  • High-performance nitrile coating reduces hand fatigue.
  • Color-coded system for easy selection.
  • Thumb reinforcement extends product life.
  • Washable for cost savings.
  • Extended cuff provides wrist protection.
  • Touchscreen compatible for operating devices without removing gloves.
  • 13 gauge for optimal fit and dexterity.
Catalog No. 50-005-08497 Supplier MSC Supplier No. 17989351
These CoreShield gloves are designed for cut, puncture, and abrasion resistance, offering a lightweight and flexible solution for hand protection. They feature an engineered yarn and high-performance nitrile coating, providing comfort and dexterity for all-day wear in various industrial applications.
  • Engineered yarn for lightweight cut protection and dexterity.
  • High-performance nitrile coating reduces hand fatigue.
  • Color-coded system for easy selection.
  • Thumb reinforcement extends product life.
  • Washable for cost savings.
  • Extended cuff provides wrist protection.
  • Touchscreen compatible for operating devices without removing gloves.
  • 13 gauge for optimal fit and dexterity.