Learn More
Biolegend Alexa Fluor 647 Annexin V
Supplier:  Biolegend 640911
Encompass_Preferred
Alexa Fluor 647 Annexin V Reactivity: All mammalian species; Apps: FC; Size: 25 tests
Catalog No. NC9032291