Learn More
Biolegend Alexa Fluor 647 anti-β-Amyloid, 1-16
Supplier:  Biolegend 803020
Encompass_Preferred
Alexa Fluor 647 anti-β-Amyloid, 1-16 [6E10]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: IHC-P; Size: 25 ug
Catalog No. 50-208-9887