Learn More
Biolegend Alexa Fluor 647 anti-β-Amyloid, 17-24
Supplier:  Biolegend 800718
Encompass_Preferred
Alexa Fluor 647 anti-β-Amyloid, 17-24 [4G8]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse; Apps: IHC-P; Size: 100 ug
Catalog No. 50-208-9828