Learn More
Biolegend Alexa Fluor 647 anti-Cytokeratin 17
Supplier:  Biolegend 697206
Encompass_Preferred
Alexa Fluor 647 anti-Cytokeratin 17 [W16131A]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: ICC; Size: 100 ug
Catalog No. 50-208-7692