Learn More
Biolegend Alexa Fluor 647 anti-human CD11a
Supplier:  Biolegend 301218
Encompass_Preferred
Alexa Fluor 647 anti-human CD11a [HI111]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cattle (Bovine, Cow), Capuchin monkey, Chimpanzee, Cynomolgus, Dog (Canine), Horse (Equine), Rabbit (Lapine), Sheep (Ovine); Apps: FC; Size: 100 tests
Catalog No. 50-165-685