Learn More
Biolegend Alexa Fluor 647 anti-human CD11c
Supplier:  Biolegend 301620
Encompass_Preferred
Alexa Fluor 647 anti-human CD11c [3.9]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Rhesus, Squirrel Monkey; Apps: FC; Size: 100 tests
Catalog No. 50-165-727