Learn More
Biolegend Alexa Fluor 647 anti-human CD127 (IL-7Rα)
Supplier:  Biolegend 351318
Encompass_Preferred
Alexa Fluor 647 anti-human CD127 (IL-7Rα) [A019D5]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-168-657